Product Details
- SNP ID
-
rs31527
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:135948376 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGATACACTAGTGGCACTGCAAACC[A/G]GTCAAGTGAGCATAAAAATAAGGCT
- Phenotype
-
MIM: 609087
MIM: 602882
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXL21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75643909] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXL21
- Gene Name
- F-box and leucine rich repeat protein 21 (gene/pseudogene)
There are no transcripts associated with this gene.
- Gene
- LECT2
- Gene Name
- leukocyte cell derived chemotaxin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002302.2 |
|
Intron |
|
|
NP_002293.2 |
View Full Product Details