Product Details

SNP ID
rs198475
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:61758599 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGGGCCTGTCGTCGGGTCAGTGAG[C/T]GTTTGTGTACATGCGTGTCCTGGCA
Phenotype
MIM: 608329
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DKFZP434K028 PubMed Links

Gene Details

Gene
DKFZP434K028
Gene Name
uncharacterized LOC26070
There are no transcripts associated with this gene.

Gene
LOC100129473
Gene Name
uncharacterized LOC100129473
There are no transcripts associated with this gene.

Gene
MYRF
Gene Name
myelin regulatory factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127392.2 Intron NP_001120864.1
NM_013279.3 Intron NP_037411.1
XM_005274222.1 Intron XP_005274279.1
XM_005274223.1 Intron XP_005274280.1
XM_005274224.1 Intron XP_005274281.1
XM_005274225.1 Intron XP_005274282.1
XM_005274226.1 Intron XP_005274283.1
XM_005274227.1 Intron XP_005274284.1
XM_005274228.1 Intron XP_005274285.1
XM_011545234.2 Intron XP_011543536.1

View Full Product Details