Product Details
- SNP ID
-
rs740427
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:89368987 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGCTCCCAGAGCTCCCCCTCCTC[C/G]CTGCCCCAAGAAGTCACGACATAAG
- Phenotype
-
MIM: 611188
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR9-3
PubMed Links
Gene Details
- Gene
- MIR9-3
- Gene Name
- microRNA 9-3
There are no transcripts associated with this gene.
- Gene
- MIR9-3HG
- Gene Name
- MIR9-3 host gene
There are no transcripts associated with this gene.
View Full Product Details