Product Details

SNP ID
rs266810
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:1397083 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGAGCAGCCGCTGGAAGTAACAGTC[A/G]CACGCTCACTGCCGACTGGCCAAGC
Phenotype
MIM: 601240 MIM: 601825
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAMT PubMed Links

Gene Details

Gene
GAMT
Gene Name
guanidinoacetate N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000156.5 1081 UTR 3 NP_000147.1
NM_138924.2 1081 Intron NP_620279.1
Gene
NDUFS7
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
There are no transcripts associated with this gene.

View Full Product Details