Product Details

SNP ID
rs204555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44426396 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAGTACAACATATGTATTATAC[A/G]TTCAGAATCCCACCACTAGAGATAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF229 PubMed Links

Gene Details

Gene
ZNF229
Gene Name
zinc finger protein 229
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278510.2 4893 UTR 3 NP_001265439.2
NM_014518.3 4893 UTR 3 NP_055333.3
XM_006723372.3 4893 UTR 3 XP_006723435.1
XM_011527292.1 4893 UTR 3 XP_011525594.1
XM_017027280.1 4893 UTR 3 XP_016882769.1

View Full Product Details