Product Details

SNP ID
rs740695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17872752 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCCGTGTGGGGGAGGCGCTGGGG[C/T]TTTGGGCCGCTGTACAGGAGGACGC
Phenotype
MIM: 604178 MIM: 601843
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL18A PubMed Links

Gene Details

Gene
RPL18A
Gene Name
ribosomal protein L18a
There are no transcripts associated with this gene.

Gene
SLC5A5
Gene Name
solute carrier family 5 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000453.2 Intron NP_000444.1
XM_011528192.2 Intron XP_011526494.1
XM_011528193.2 Intron XP_011526495.1
XM_011528194.2 Intron XP_011526496.1
XM_017027158.1 Intron XP_016882647.1

View Full Product Details