Product Details

SNP ID
rs42507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93188980 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGAGATATTTCATACAAAACTTA[C/T]GAGGAAACCCCTGTCACTTTCGTTC
Phenotype
MIM: 614133
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HEPACAM2 PubMed Links

Gene Details

Gene
HEPACAM2
Gene Name
HEPACAM family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039372.2 1741 UTR 3 NP_001034461.1
NM_001288804.1 1741 UTR 3 NP_001275733.1
NM_001288810.1 1741 UTR 3 NP_001275739.1
NM_198151.2 1741 UTR 3 NP_937794.1
XM_011516000.2 1741 UTR 3 XP_011514302.1
XM_011516001.1 1741 UTR 3 XP_011514303.1

View Full Product Details