Product Details

SNP ID
rs368421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63848114 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACACTTCCTGCAATAGCACTAGG[A/G]AGTCTTTCTCAGGGAGCAAGGCCAG
Phenotype
MIM: 600526
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARK2 PubMed Links

Gene Details

Gene
MARK2
Gene Name
microtubule affinity regulating kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039469.2 Intron NP_001034558.2
NM_001163296.1 Intron NP_001156768.1
NM_001163297.1 Intron NP_001156769.1
NM_004954.4 Intron NP_004945.4
NM_017490.3 Intron NP_059672.2
XM_011544789.1 Intron XP_011543091.1
XM_011544790.1 Intron XP_011543092.1
XM_011544791.1 Intron XP_011543093.1
XM_011544792.1 Intron XP_011543094.1
XM_011544793.1 Intron XP_011543095.1
XM_011544794.1 Intron XP_011543096.1
XM_011544795.1 Intron XP_011543097.1
XM_011544796.1 Intron XP_011543098.1
XM_011544797.1 Intron XP_011543099.1
XM_011544798.1 Intron XP_011543100.1
XM_017017310.1 Intron XP_016872799.1
XM_017017311.1 Intron XP_016872800.1

View Full Product Details