Product Details

SNP ID
rs128624216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153725709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGCCCTCCCTGCTACCTTCGTCA[A/G]CAGTGCCATCCGTTACCTGGAGGGC
Phenotype
MIM: 300371 MIM: 300398
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCD1 PubMed Links

Gene Details

Gene
ABCD1
Gene Name
ATP binding cassette subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000033.3 842 Missense Mutation AAC,AGC N148S NP_000024.2
Gene
BCAP31
Gene Name
B-cell receptor-associated protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139441.1 842 Intron NP_001132913.1
NM_001139457.2 842 Intron NP_001132929.1
NM_001256447.1 842 Intron NP_001243376.1
NM_005745.7 842 Intron NP_005736.3

View Full Product Details