Product Details

SNP ID
rs363156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66758811 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAGTAGATTCATTACTGAGTCGT[C/T]GTCTTCTACGTGGGGTCAGCAGAAT
Phenotype
MIM: 611406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYNC1LI2 PubMed Links

Gene Details

Gene
DYNC1LI2
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
There are no transcripts associated with this gene.

Gene
LOC106699570
Gene Name
uncharacterized LOC106699570
There are no transcripts associated with this gene.

Gene
TERB1
Gene Name
telomere repeat binding bouquet formation protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136505.1 2220 Missense Mutation NP_001129977.1
XM_011523004.2 2220 Missense Mutation XP_011521306.1
XM_011523005.2 2220 Missense Mutation XP_011521307.1
XM_011523006.2 2220 Missense Mutation XP_011521308.1
XM_011523007.2 2220 Missense Mutation XP_011521309.1
XM_011523008.2 2220 Missense Mutation XP_011521310.1
XM_011523009.2 2220 Missense Mutation XP_011521311.1
XM_011523010.2 2220 Missense Mutation XP_011521312.1
XM_011523011.2 2220 Missense Mutation XP_011521313.1
XM_011523012.2 2220 Missense Mutation XP_011521314.1
XM_011523014.2 2220 Missense Mutation XP_011521316.1
XM_011523015.2 2220 Missense Mutation XP_011521317.1
XM_011523016.2 2220 Missense Mutation XP_011521318.1
XM_011523017.1 2220 UTR 3 XP_011521319.1
XM_011523018.1 2220 Intron XP_011521320.1
XM_011523020.1 2220 Intron XP_011521322.1
XM_017023157.1 2220 UTR 3 XP_016878646.1
XM_017023158.1 2220 UTR 3 XP_016878647.1
XM_017023159.1 2220 UTR 3 XP_016878648.1

View Full Product Details