Product Details

SNP ID
rs363203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66805747 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAGTAACTGTACTATGTAACAA[A/C]AGGTGAGTCTTCTCATATATGGAAC
Phenotype
MIM: 603385
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NAE1 PubMed Links

Gene Details

Gene
NAE1
Gene Name
NEDD8 activating enzyme E1 subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018159.1 Intron NP_001018169.1
NM_001018160.1 Intron NP_001018170.1
NM_001286500.1 Intron NP_001273429.1
NM_003905.3 Intron NP_003896.1
XM_005256215.1 Intron XP_005256272.1
XM_011523422.2 Intron XP_011521724.1
Gene
TERB1
Gene Name
telomere repeat binding bouquet formation protein 1
There are no transcripts associated with this gene.

View Full Product Details