Product Details

SNP ID
rs6656
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:44566499 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTTCTTCCGCTTCTTGTGAGGGCG[C/T]ACCAGGCCACGGAGAGCAGGAGGAA
Phenotype
MIM: 608023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX56 PubMed Links

Gene Details

Gene
DDX56
Gene Name
DEAD-box helicase 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257189.1 2038 Silent Mutation GTA,GTG V465V NP_001244118.1
NM_019082.3 2038 Silent Mutation GTA,GTG V505V NP_061955.1

View Full Product Details