Product Details

SNP ID
rs212429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:105877273 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAACGCAGGTGAGTTTGGAGCGC[G/T]ACGAGCTACCTTCCCATCTCAGGCT
Phenotype
MIM: 615610
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATXN7L1 PubMed Links

Gene Details

Gene
ATXN7L1
Gene Name
ataxin 7 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318229.1 Intron NP_001305158.1
NM_020725.1 Intron NP_065776.1
NM_138495.1 Intron NP_612504.1
NM_152749.3 Intron NP_689962.1
XM_005250220.3 Intron XP_005250277.1
XM_006715900.2 Intron XP_006715963.1
XM_006715901.2 Intron XP_006715964.1
XM_006715902.3 Intron XP_006715965.1
XM_006715903.3 Intron XP_006715966.1
XM_006715904.3 Intron XP_006715967.1
XM_011515935.1 Intron XP_011514237.1
XM_011515936.2 Intron XP_011514238.1
XM_011515937.2 Intron XP_011514239.1
XM_011515938.2 Intron XP_011514240.1
XM_011515939.1 Intron XP_011514241.1
XM_011515940.1 Intron XP_011514242.1
XM_011515941.2 Intron XP_011514243.1
XM_011515942.2 Intron XP_011514244.1
XM_011515943.2 Intron XP_011514245.1
XM_011515944.1 Intron XP_011514246.1
XM_011515945.1 Intron XP_011514247.1
XM_011515946.1 Intron XP_011514248.1
XM_011515947.1 Intron XP_011514249.1
XM_011515950.1 Intron XP_011514252.1
XM_011515954.2 Intron XP_011514256.1
XM_017011858.1 Intron XP_016867347.1
XM_017011859.1 Intron XP_016867348.1
XM_017011860.1 Intron XP_016867349.1
XM_017011861.1 Intron XP_016867350.1
Gene
CDHR3
Gene Name
cadherin related family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301161.1 Intron NP_001288090.1
NM_152750.4 Intron NP_689963.2
XM_005250224.4 Intron XP_005250281.1
XM_005250225.2 Intron XP_005250282.1
XM_006715905.2 Intron XP_006715968.1
XM_011515955.2 Intron XP_011514257.1
XM_011515956.2 Intron XP_011514258.1
XM_011515957.2 Intron XP_011514259.1
XM_011515958.2 Intron XP_011514260.1
XM_011515959.2 Intron XP_011514261.1
XM_017011862.1 Intron XP_016867351.1
XM_017011863.1 Intron XP_016867352.1
XM_017011864.1 Intron XP_016867353.1
XM_017011865.1 Intron XP_016867354.1
XM_017011866.1 Intron XP_016867355.1
XM_017011867.1 Intron XP_016867356.1

View Full Product Details