Product Details

SNP ID
hCV2295217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30304595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATGACTTTATATCTTTGTACAG[A/C]GCATTTCCAGCATCATATCATCCAT
Phenotype
MIM: 300473
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NR0B1 PubMed Links

Gene Details

Gene
NR0B1
Gene Name
nuclear receptor subfamily 0 group B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000475.4 1412 Missense Mutation CGC,CTC R466L NP_000466.2

View Full Product Details