Product Details

SNP ID
rs763995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:108150200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATTGGCATTTAGGTAACATTTT[C/G]TGTTTTTAAAGACCATGCATGCAAA
Phenotype
MIM: 601581
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NRCAM PubMed Links

Gene Details

Gene
NRCAM
Gene Name
neuronal cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037132.2 Intron NP_001032209.1
NM_001193582.1 Intron NP_001180511.1
NM_001193583.1 Intron NP_001180512.1
NM_001193584.1 Intron NP_001180513.1
NM_005010.4 Intron NP_005001.3
XM_005250373.2 Intron XP_005250430.1
XM_005250383.2 Intron XP_005250440.1
XM_005250385.2 Intron XP_005250442.1
XM_006716003.1 Intron XP_006716066.1
XM_006716007.1 Intron XP_006716070.1
XM_006716012.1 Intron XP_006716075.1
XM_006716014.1 Intron XP_006716077.1
XM_011516253.1 Intron XP_011514555.1
XM_011516255.1 Intron XP_011514557.1
XM_011516256.1 Intron XP_011514558.1
XM_011516257.1 Intron XP_011514559.1
XM_011516258.1 Intron XP_011514560.1
XM_011516259.1 Intron XP_011514561.1
XM_011516261.1 Intron XP_011514563.1
XM_011516262.1 Intron XP_011514564.1
XM_011516265.2 Intron XP_011514567.1
XM_011516266.2 Intron XP_011514568.1
XM_011516267.1 Intron XP_011514569.1
XM_011516268.2 Intron XP_011514570.1
XM_011516269.1 Intron XP_011514571.1
XM_011516270.1 Intron XP_011514572.1
XM_011516271.1 Intron XP_011514573.1
XM_017012236.1 Intron XP_016867725.1
XM_017012237.1 Intron XP_016867726.1
XM_017012238.1 Intron XP_016867727.1
XM_017012239.1 Intron XP_016867728.1
XM_017012240.1 Intron XP_016867729.1
XM_017012241.1 Intron XP_016867730.1
XM_017012242.1 Intron XP_016867731.1
XM_017012243.1 Intron XP_016867732.1
XM_017012244.1 Intron XP_016867733.1
XM_017012245.1 Intron XP_016867734.1
XM_017012246.1 Intron XP_016867735.1
XM_017012247.1 Intron XP_016867736.1
XM_017012248.1 Intron XP_016867737.1
XM_017012249.1 Intron XP_016867738.1
XM_017012250.1 Intron XP_016867739.1
XM_017012251.1 Intron XP_016867740.1
XM_017012252.1 Intron XP_016867741.1
XM_017012253.1 Intron XP_016867742.1
XM_017012254.1 Intron XP_016867743.1
XM_017012255.1 Intron XP_016867744.1
XM_017012256.1 Intron XP_016867745.1
XM_017012257.1 Intron XP_016867746.1
XM_017012258.1 Intron XP_016867747.1
XM_017012259.1 Intron XP_016867748.1

View Full Product Details