Product Details

SNP ID
rs500135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18026098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACACTTTCCTCCATCCCCCTTT[C/T]TCAGCTTTATTTTTCTCCATGGGAC
Phenotype
MIM: 191060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPH1 PubMed Links

Gene Details

Gene
TPH1
Gene Name
tryptophan hydroxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004179.2 Intron NP_004170.1

View Full Product Details