Product Details

SNP ID
rs41408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:72183874 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATACTTGGCCTGGCTTGTTGGGGG[C/T]TCAGGAATTAATAAGCCACAGGAGG
Phenotype
MIM: 607880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOC6B PubMed Links

Gene Details

Gene
EXOC6B
Gene Name
exocyst complex component 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321729.1 Intron NP_001308658.1
NM_001321730.1 Intron NP_001308659.1
NM_001321731.1 Intron NP_001308660.1
NM_001321733.1 Intron NP_001308662.1
NM_001321734.1 Intron NP_001308663.1
NM_015189.2 Intron NP_056004.1
XM_005264224.1 Intron XP_005264281.1
XM_011532711.2 Intron XP_011531013.1
XM_011532712.2 Intron XP_011531014.1
XM_017003641.1 Intron XP_016859130.1
XM_017003642.1 Intron XP_016859131.1

View Full Product Details