Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001321729.1 | Intron | NP_001308658.1 | |||
NM_001321730.1 | Intron | NP_001308659.1 | |||
NM_001321731.1 | Intron | NP_001308660.1 | |||
NM_001321733.1 | Intron | NP_001308662.1 | |||
NM_001321734.1 | Intron | NP_001308663.1 | |||
NM_015189.2 | Intron | NP_056004.1 | |||
XM_005264224.1 | Intron | XP_005264281.1 | |||
XM_011532711.2 | Intron | XP_011531013.1 | |||
XM_011532712.2 | Intron | XP_011531014.1 | |||
XM_017003641.1 | Intron | XP_016859130.1 | |||
XM_017003642.1 | Intron | XP_016859131.1 |