Product Details

SNP ID
rs41414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:72178698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCTGTATCCTGCCCTAGAAAAA[A/T]TTGCAATAAAAGCCTGGGGTTAGAT
Phenotype
MIM: 607880
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EXOC6B PubMed Links

Gene Details

Gene
EXOC6B
Gene Name
exocyst complex component 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321729.1 2509 UTR 3 NP_001308658.1
NM_001321730.1 2509 UTR 3 NP_001308659.1
NM_001321731.1 2509 UTR 3 NP_001308660.1
NM_001321733.1 2509 UTR 3 NP_001308662.1
NM_001321734.1 2509 UTR 3 NP_001308663.1
NM_015189.2 2509 UTR 3 NP_056004.1
XM_005264224.1 2509 UTR 3 XP_005264281.1
XM_011532711.2 2509 Intron XP_011531013.1
XM_011532712.2 2509 Intron XP_011531014.1
XM_017003641.1 2509 UTR 3 XP_016859130.1
XM_017003642.1 2509 UTR 3 XP_016859131.1

View Full Product Details