Product Details

SNP ID
rs741432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12934856 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGAAGGAGGTCCCCCAGCAGGT[C/G]CTTGTGCCCGCGGGTTGTTTGTGAT
Phenotype
MIM: 607437
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM234B PubMed Links

Gene Details

Gene
FAM234B
Gene Name
family with sequence similarity 234 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020853.1 Intron NP_065904.1
XM_017019706.1 Intron XP_016875195.1
Gene
GPRC5D
Gene Name
G protein-coupled receptor class C group 5 member D
There are no transcripts associated with this gene.

View Full Product Details