Product Details

SNP ID
rs363711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74655835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAGTACATCTAGGAATGAATGG[G/T]TTGGTGTCCAGTCTGCAGTCTTGAG
Phenotype
MIM: 603706
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
M1AP PubMed Links

Gene Details

Gene
M1AP
Gene Name
meiosis 1 associated protein
There are no transcripts associated with this gene.

Gene
SEMA4F
Gene Name
ssemaphorin 4F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271661.1 Intron NP_001258590.1
NM_001271662.1 Intron NP_001258591.1
NM_004263.4 Intron NP_004254.2
XM_011532475.2 Intron XP_011530777.1
XM_011532476.2 Intron XP_011530778.1
XM_011532477.2 Intron XP_011530779.1
XM_017003172.1 Intron XP_016858661.1
XM_017003173.1 Intron XP_016858662.1
XM_017003174.1 Intron XP_016858663.1

View Full Product Details