Product Details
- SNP ID
-
rs768049
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:68202331 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGTGTTAGCAACTCTCGAAACT[A/C]TATGACAGGGATTTATAGGAACCGC
- Phenotype
-
MIM: 605679
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL26
PubMed Links
Gene Details
- Gene
- IL26
- Gene Name
- interleukin 26
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018402.1 |
|
Intron |
|
|
NP_060872.1 |
View Full Product Details