Product Details

SNP ID
rs369570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:93229403 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCCAACATGATTTCAGATAAGAT[A/G]TCTCAAAGAAGAAAATAGTTAGGTA
Phenotype
MIM: 604404
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPC6 PubMed Links

Gene Details

Gene
GPC6
Gene Name
glypican 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005708.3 Intron NP_005699.1
XM_011521044.2 Intron XP_011519346.1
XM_017020298.1 Intron XP_016875787.1
XM_017020299.1 Intron XP_016875788.1
XM_017020300.1 Intron XP_016875789.1
XM_017020301.1 Intron XP_016875790.1
XM_017020302.1 Intron XP_016875791.1

View Full Product Details