Product Details

SNP ID
rs334877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55222859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGTCGTTGACAGTTGGCTGGGTT[C/T]ACCACCTTCACGACCACGAAATCTG
Phenotype
MIM: 607752 MIM: 614086
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNO PubMed Links

Gene Details

Gene
CCNO
Gene Name
cyclin O
There are no transcripts associated with this gene.

Gene
MCIDAS
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190787.1 312 Intron NP_001177716.1
XM_017009439.1 312 UTR 5 XP_016864928.1

View Full Product Details