Product Details

SNP ID
rs461103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55219650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTATAGGTGGTTATTCTATTTGCA[A/C]AAATAGAACTGTATGGATAATGAAT
Phenotype
MIM: 614086
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MCIDAS PubMed Links

Gene Details

Gene
MCIDAS
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190787.1 2051 UTR 3 NP_001177716.1
XM_017009439.1 2051 Intron XP_016864928.1

View Full Product Details