Product Details

SNP ID
rs459724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55218643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTAGTCCACAAGATGACGCCCC[A/G]ACCTCCAACCTCCCACCCCAACCCC
Phenotype
MIM: 614086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCIDAS PubMed Links

Gene Details

Gene
MCIDAS
Gene Name
multiciliate differentiation and DNA synthesis associated cell cycle protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190787.1 Intron NP_001177716.1
XM_017009439.1 Intron XP_016864928.1

View Full Product Details