Product Details

SNP ID
rs10412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33110841 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTCATCCTGGCTATGCAGAGCCCC[C/T]GCCCCTGGCAGTCAGCAGAGACCCT
Phenotype
MIM: 137060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B4GALT1 PubMed Links

Gene Details

Gene
B4GALT1
Gene Name
beta-1,4-galactosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001497.3 3909 UTR 3 NP_001488.2
XM_005251440.4 3909 UTR 3 XP_005251497.1

View Full Product Details