Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002220.2 | 3130 | Intron | NP_002211.1 | ||
XM_011521522.2 | 3130 | Intron | XP_011519824.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135685.1 | 3130 | UTR 3 | NP_001129157.1 | ||
NM_002344.5 | 3130 | UTR 3 | NP_002335.2 | ||
NM_206961.3 | 3130 | UTR 3 | NP_996844.1 | ||
XM_011521556.2 | 3130 | Intron | XP_011519858.1 | ||
XM_011521557.2 | 3130 | Intron | XP_011519859.1 | ||
XM_017022181.1 | 3130 | UTR 3 | XP_016877670.1 | ||
XM_017022182.1 | 3130 | Intron | XP_016877671.1 | ||
XM_017022183.1 | 3130 | Intron | XP_016877672.1 |