Product Details

SNP ID
rs767844
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99740715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCCCTCCTTTAGAGACTTAAGCT[A/G]TTCTTACATTAGACTACTTGAAATT
Phenotype
MIM: 603646 MIM: 610101
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
COX15 PubMed Links

Gene Details

Gene
COX15
Gene Name
COX15, cytochrome c oxidase assembly homolog
There are no transcripts associated with this gene.

Gene
CUTC
Gene Name
cutC copper transporter
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015960.2 Intron NP_057044.2

View Full Product Details