Product Details
- SNP ID
-
hCV2380159
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:100976078 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGGCCTGAGCAGGCTTATGGAGA[C/G]GGGGGACAGATGGTAGTGGCTTGTA
- Phenotype
-
MIM: 611848
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR608
PubMed Links
Gene Details
- Gene
- MIR608
- Gene Name
- microRNA 608
There are no transcripts associated with this gene.
- Gene
- MRPL43
- Gene Name
- mitochondrial ribosomal protein L43
There are no transcripts associated with this gene.
- Gene
- SEMA4G
- Gene Name
- semaphorin 4G
There are no transcripts associated with this gene.
View Full Product Details