Product Details
- SNP ID
-
rs663849
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:139042256 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAGCCCAAAGCCTGGTCTCATTA[A/G]CGCTCTTGCCAGGTTTCTGTACGTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRR23C
PubMed Links
Gene Details
- Gene
- PRR23C
- Gene Name
- proline rich 23C
View Full Product Details