Product Details

SNP ID
rs250584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23391710 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTAGGCTAAAGAGCAGACAAGGTA[A/T]TGGAATGGCATCAAGAGACTGGGGA
Phenotype
MIM: 606978
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COG7 PubMed Links

Gene Details

Gene
COG7
Gene Name
component of oligomeric golgi complex 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153603.3 Intron NP_705831.1
XM_017023870.1 Intron XP_016879359.1

View Full Product Details