Product Details
- SNP ID
-
rs33673
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:36153735 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAATTCCAACCTTCTTCTCCCCAG[A/G]TCTATCCCCCGAACTAGAATGGATT
- Phenotype
-
MIM: 601436
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LMBRD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75356410] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LMBRD2
- Gene Name
- LMBR1 domain containing 2
There are no transcripts associated with this gene.
- Gene
- MIR580
- Gene Name
- microRNA 580
There are no transcripts associated with this gene.
- Gene
- SKP2
- Gene Name
- S-phase kinase-associated protein 2, E3 ubiquitin protein ligase
View Full Product Details