Product Details

SNP ID
rs152053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132191043 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCCGAGATCAAGATGCTAGTAGG[C/T]TTGTTTCTGGTAAGGGCCCCCTGGT
Phenotype
MIM: 600608
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P4HA2 PubMed Links

Gene Details

Gene
P4HA2
Gene Name
prolyl 4-hydroxylase subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017973.1 3484 Intron NP_001017973.1
NM_001017974.1 3484 Intron NP_001017974.1
NM_001142598.1 3484 Intron NP_001136070.1
NM_001142599.1 3484 Intron NP_001136071.1
NM_004199.2 3484 Intron NP_004190.1
XM_005272116.4 3484 UTR 3 XP_005272173.1
XM_005272117.4 3484 UTR 3 XP_005272174.1
XM_005272118.4 3484 UTR 3 XP_005272175.1
XM_005272119.4 3484 UTR 3 XP_005272176.1
XM_005272120.4 3484 UTR 3 XP_005272177.1
XM_006714728.3 3484 UTR 3 XP_006714791.1
XM_006714729.3 3484 UTR 3 XP_006714792.1
XM_006714730.3 3484 UTR 3 XP_006714793.1
XM_011543705.2 3484 UTR 3 XP_011542007.1
XM_017010011.1 3484 UTR 3 XP_016865500.1
XM_017010012.1 3484 UTR 3 XP_016865501.1
Gene
P4HA2-AS1
Gene Name
P4HA2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details