Product Details

SNP ID
rs34323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12726123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAACTGGGGAGTGCGGGAGGGTG[A/G]AGGTGGCCCGGAAAAGAACACCTCT
Phenotype
MIM: 612456 MIM: 600778 MIM: 602927
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOLD1 PubMed Links

Gene Details

Gene
APOLD1
Gene Name
apolipoprotein L domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130415.1 Intron NP_001123887.1
NM_030817.2 Intron NP_110444.3
Gene
CDKN1B
Gene Name
cyclin dependent kinase inhibitor 1B
There are no transcripts associated with this gene.

Gene
GPR19
Gene Name
G protein-coupled receptor 19
There are no transcripts associated with this gene.

View Full Product Details