Product Details

SNP ID
rs279254
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11942504 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAAAACCTCTCAGACACTGGGTT[A/G]GAGAAGGAAGTGGCTTTATTCGGCC
Phenotype
MIM: 194543
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF69 PubMed Links
Additional Information
For this assay, SNP(s) [rs113531904] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ZNF69
Gene Name
zinc finger protein 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321163.1 Intron NP_001308092.1
NM_021915.3 Intron NP_068734.1
XM_006722873.3 Intron XP_006722936.1
XM_006722874.3 Intron XP_006722937.1
XM_017027231.1 Intron XP_016882720.1
Gene
ZNF700
Gene Name
zinc finger protein 700
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271848.1 Intron NP_001258777.1
NM_144566.2 Intron NP_653167.1

View Full Product Details