Product Details

SNP ID
rs505296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:36489068 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTGAATAGACAAAAAGGGTTACA[C/T]AGCATAACAATGAGCAGTTGGTATC
Phenotype
MIM: 602355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAF6 PubMed Links

Gene Details

Gene
TRAF6
Gene Name
TNF receptor associated factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004620.3 2627 UTR 3 NP_004611.1
NM_145803.2 2627 UTR 3 NP_665802.1
XM_017018220.1 2627 Intron XP_016873709.1

View Full Product Details