Product Details

SNP ID
rs549377
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:73320492 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATGTGGGACCAGCCCTTCTCATTA[C/T]TTGGGGATTGCAGCCATACCTGCTC
Phenotype
MIM: 605206
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCN4 PubMed Links

Gene Details

Gene
HCN4
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005477.2 6595 UTR 3 NP_005468.1
XM_011521148.2 6595 UTR 3 XP_011519450.1

View Full Product Details