Product Details

SNP ID
rs279941
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.13:103045818 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
CATAAGAATTCAGTTTTGGTGTTAA[A/C]GATGTTTTCATTCTCGGTGTTCCCT
Phenotype
MIM: 601295
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC10A2 PubMed Links

Gene Details

Gene
SLC10A2
Gene Name
solute carrier family 10 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000452.2 1959 UTR 3 NP_000443.1

View Full Product Details