Product Details

SNP ID
rs254255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54105871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATCTTCCCTTTGCCAAGGCTCA[C/G]CTTCTCTTCCCCTCTCTTCAGAGCC
Phenotype
MIM: 603832 MIM: 606862 MIM: 606419 MIM: 609519
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NDUFA3 PubMed Links

Gene Details

Gene
NDUFA3
Gene Name
NADH:ubiquinone oxidoreductase subunit A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004542.3 Intron NP_004533.1
XM_017026833.1 Intron XP_016882322.1
Gene
OSCAR
Gene Name
osteoclast associated, immunoglobulin-like receptor
There are no transcripts associated with this gene.

Gene
PRPF31
Gene Name
pre-mRNA processing factor 31
There are no transcripts associated with this gene.

Gene
TFPT
Gene Name
TCF3 fusion partner
There are no transcripts associated with this gene.

View Full Product Details