Product Details

SNP ID
rs254278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54120396 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTACTGGAGGTGCACAAGTGCTG[C/T]GGGTACACAGAGGAGGCGTCTGATC
Phenotype
MIM: 606419 MIM: 609519
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRPF31 PubMed Links

Gene Details

Gene
PRPF31
Gene Name
pre-mRNA processing factor 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015629.3 Intron NP_056444.3
XM_006723137.3 Intron XP_006723200.1
Gene
TFPT
Gene Name
TCF3 fusion partner
There are no transcripts associated with this gene.

View Full Product Details