Product Details

SNP ID
rs357509
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:88192125 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGAGATGTTTTCTGATTCTTAGA[C/T]TAGCATTCTGCCTACGAAACTATGA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TMEM161B PubMed Links

Gene Details

Gene
TMEM161B
Gene Name
transmembrane protein 161B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289007.1 Intron NP_001275936.1
NM_001289008.1 Intron NP_001275937.1
NM_153354.4 Intron NP_699185.1
XM_006714553.1 Intron XP_006714616.1
XM_006714554.3 Intron XP_006714617.1
XM_006714555.3 Intron XP_006714618.1
XM_006714556.3 Intron XP_006714619.2
XM_011543201.2 Intron XP_011541503.1
XM_011543202.2 Intron XP_011541504.2
XM_011543203.2 Intron XP_011541505.2
XM_011543204.2 Intron XP_011541506.2
XM_017009093.1 Intron XP_016864582.1
XM_017009094.1 Intron XP_016864583.1
XM_017009095.1 Intron XP_016864584.1
XM_017009096.1 Intron XP_016864585.1
XM_017009097.1 Intron XP_016864586.1
XM_017009098.1 Intron XP_016864587.1
XM_017009099.1 Intron XP_016864588.1
XM_017009100.1 Intron XP_016864589.1
XM_017009101.1 Intron XP_016864590.1
XM_017009102.1 Intron XP_016864591.1
XM_017009103.1 Intron XP_016864592.1
XM_017009104.1 Intron XP_016864593.1
XM_017009105.1 Intron XP_016864594.1
XM_017009106.1 Intron XP_016864595.1
XM_017009107.1 Intron XP_016864596.1
XM_017009108.1 Intron XP_016864597.1
XM_017009109.1 Intron XP_016864598.1
XM_017009110.1 Intron XP_016864599.1

View Full Product Details