Product Details

SNP ID
rs367674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:23305615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCAATTAGTTTAGCCCAGGTGA[C/G]TCCAGTCAGCCGATATTTGAGCCCA
Phenotype
MIM: 603971
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF91 PubMed Links

Gene Details

Gene
ZNF91
Gene Name
zinc finger protein 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300951.1 Intron NP_001287880.1
NM_003430.3 Intron NP_003421.2
XM_006722880.3 Intron XP_006722943.1
XM_011528265.2 Intron XP_011526567.1
XM_011528266.2 Intron XP_011526568.1
XM_017027235.1 Intron XP_016882724.1
XM_017027236.1 Intron XP_016882725.1

View Full Product Details