Product Details

SNP ID
rs167707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52336691 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGGCGGGGGAGGCCGCGTCCTGT[A/G]CTGGGTCCTGGGACCCTTGAGACCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF610 PubMed Links

Gene Details

Gene
ZNF610
Gene Name
zinc finger protein 610
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161425.1 Intron NP_001154897.1
NM_001161426.1 Intron NP_001154898.1
NM_001161427.1 Intron NP_001154899.1
NM_173530.2 Intron NP_775801.2

View Full Product Details