Product Details

SNP ID
rs9603791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:40797964 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTAGGGTGGTTACAGTTCATTCT[C/T]CTCTTCTGTTTAGAATTTTTACTAA
Phenotype
MIM: 603861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A15 PubMed Links

Gene Details

Gene
SLC25A15
Gene Name
solute carrier family 25 member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014252.3 Intron NP_055067.1
Gene
TPTE2P5
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 5
There are no transcripts associated with this gene.

View Full Product Details