Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001013732.3 | 20705 | Intron | NP_001013754.3 | ||
NM_207499.2 | 20705 | Intron | NP_997382.2 | ||
XM_011514639.2 | 20705 | UTR 3 | XP_011512941.1 | ||
XM_017010891.1 | 20705 | UTR 3 | XP_016866380.1 | ||
XM_017010892.1 | 20705 | UTR 3 | XP_016866381.1 | ||
XM_017010893.1 | 20705 | UTR 3 | XP_016866382.1 | ||
XM_017010894.1 | 20705 | UTR 3 | XP_016866383.1 | ||
XM_017010895.1 | 20705 | UTR 3 | XP_016866384.1 | ||
XM_017010896.1 | 20705 | Intron | XP_016866385.1 |