Product Details

SNP ID
rs3748346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21032936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCCAATGGTTACATCTTACA[C/T]ACTCAGATGTGGTTTTAGAAATTTA
Phenotype
MIM: 605272 MIM: 612484 MIM: 616956
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6717 PubMed Links

Gene Details

Gene
MIR6717
Gene Name
microRNA 6717
There are no transcripts associated with this gene.

Gene
NDRG2
Gene Name
NDRG family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282211.1 1491 Intron NP_001269140.1
NM_001282212.1 1491 Intron NP_001269141.1
NM_001282213.1 1491 Intron NP_001269142.1
NM_001282214.1 1491 Intron NP_001269143.1
NM_001282215.1 1491 Intron NP_001269144.1
NM_001282216.1 1491 Intron NP_001269145.1
NM_001320329.1 1491 Intron NP_001307258.1
NM_016250.2 1491 Intron NP_057334.1
NM_201535.1 1491 Intron NP_963293.1
NM_201536.1 1491 Intron NP_963294.1
NM_201537.1 1491 Intron NP_963831.1
NM_201538.1 1491 Intron NP_963832.1
NM_201539.1 1491 Intron NP_963833.1
NM_201540.1 1491 Intron NP_963834.1
NM_201541.1 1491 Intron NP_963835.1
XM_011536996.2 1491 Intron XP_011535298.1
XM_011536997.1 1491 Intron XP_011535299.1
XM_011536998.1 1491 Intron XP_011535300.1
XM_011536999.1 1491 Intron XP_011535301.1
XM_011537001.1 1491 Intron XP_011535303.1
XM_011537002.1 1491 Intron XP_011535304.1
XM_017021480.1 1491 Intron XP_016876969.1
XM_017021481.1 1491 Intron XP_016876970.1
XM_017021482.1 1491 Intron XP_016876971.1
XM_017021483.1 1491 Intron XP_016876972.1
XM_017021484.1 1491 Intron XP_016876973.1
XM_017021485.1 1491 Intron XP_016876974.1
XM_017021486.1 1491 Intron XP_016876975.1
XM_017021487.1 1491 Intron XP_016876976.1
XM_017021488.1 1491 Intron XP_016876977.1
XM_017021489.1 1491 Intron XP_016876978.1
XM_017021490.1 1491 Intron XP_016876979.1
XM_017021491.1 1491 Intron XP_016876980.1
XM_017021492.1 1491 Intron XP_016876981.1
XM_017021493.1 1491 Intron XP_016876982.1
XM_017021494.1 1491 Intron XP_016876983.1
XM_017021495.1 1491 Intron XP_016876984.1
XM_017021496.1 1491 Intron XP_016876985.1
XM_017021497.1 1491 Intron XP_016876986.1
Gene
RNASE13
Gene Name
ribonuclease A family member 13 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012264.3 1491 UTR 3 NP_001012264.1
Gene
RNASE7
Gene Name
ribonuclease A family member 7
There are no transcripts associated with this gene.

Gene
TPPP2
Gene Name
tubulin polymerization promoting protein family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173846.4 1491 Intron NP_776245.2
XM_005267324.4 1491 Intron XP_005267381.1
XM_011536416.1 1491 Intron XP_011534718.1
XM_011536417.2 1491 Intron XP_011534719.1
XM_011536418.1 1491 Intron XP_011534720.1
XM_011536420.2 1491 Intron XP_011534722.1
XM_017020966.1 1491 Intron XP_016876455.1

View Full Product Details