Product Details
- SNP ID
-
rs1158632
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:63787744 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGGACCTCGGAGAGGACAAAGTGC[C/T]CTGGGTCCATAGGCAGCCATGGGCT
- Phenotype
-
MIM: 608442
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SYNE2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141720811] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SYNE2
- Gene Name
- spectrin repeat containing nuclear envelope protein 2
View Full Product Details