Product Details

SNP ID
rs11760149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30618411 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGGGATCCCAATTTCTACCAACC[C/T]TGCTATTTCTCAATAAGGTCAGGAT
Phenotype
MIM: 615556 MIM: 611982 MIM: 603771
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATAT1 PubMed Links

Gene Details

Gene
ATAT1
Gene Name
alpha tubulin acetyltransferase 1
There are no transcripts associated with this gene.

Gene
MRPS18B
Gene Name
mitochondrial ribosomal protein S18B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014046.3 Intron NP_054765.1
Gene
PPP1R10
Gene Name
protein phosphatase 1 regulatory subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002714.3 Intron NP_002705.2
XM_006715130.2 Intron XP_006715193.1
XM_011514722.1 Intron XP_011513024.1
XM_017010995.1 Intron XP_016866484.1

View Full Product Details