Product Details

SNP ID
rs1116221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30103553 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCTGGCTTAGCTTGAAGGAACCT[C/T]ACAAAACCAAGCCCGGATCGCTGTC
Phenotype
MIM: 609316
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM31 PubMed Links

Gene Details

Gene
TRIM31
Gene Name
tripartite motif containing 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007028.4 1387 Missense Mutation AAG,GAG K421E NP_008959.3
XM_011514264.1 1387 Missense Mutation AAG,GAG K420E XP_011512566.1
XM_011514265.1 1387 Intron XP_011512567.1
XM_011514266.1 1387 Intron XP_011512568.1
Gene
TRIM31-AS1
Gene Name
TRIM31 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details